Please login to your online account to display your discounted pricing

4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
CAS 220462-27-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7F3N2O2
Formula Weight 256.18
MDL Number MFCD03086203
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00070DA

  SDS
fisher scientific company
MO00070DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO00070CB

  SDS
fisher scientific company
MO00070CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.18
Quantity 1g
Assay 97%
CAS Min % 97.0
CAS 220462-27-1
Chemical Name or Material 4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Molecular Formula C11H7F3N2O2
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03086203
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.