Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Pyridyl)benzaldehyde, 98%

$151.76 - $151.76

Chemical Identifiers

Chemical Name or Material 4-(3-Pyridyl)benzaldehyde
CAS 127406-55-7
Molecular Formula C12H9NO
Formula Weight 183.21g/mol
MDL Number MFCD01631910
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107703

 
alfa aesar™
H51077-03
Each for $151.76
Description & Specifications

Specifications

Melting Point 52° to 57°C
Quantity 1g
Chemical Name or Material 4-(3-Pyridyl)benzaldehyde
CAS 127406-55-7
Assay 98%
Molecular Formula C12H9NO
Formula Weight 183.21g/mol
MDL Number MFCD01631910
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.