missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Pyridyl)-2-thiazoleacetonitrile, 97%

Chemical Identifiers

CAS 1016859-62-3
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.247
InChI Key FAUDLOFDWKFOQO-UHFFFAOYSA-N
Synonym 4-3-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-3-yl thiazol-2-yl acetonitrile, 2-4-pyridin-3-yl-1,3-thiazol-2-yl acetonitrile
PubChem CID 24704442
IUPAC Name 2-(4-pyridin-3-yl-1,3-thiazol-2-yl)acetonitrile
SMILES C1=CC(=CN=C1)C2=CSC(=N2)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66267MD View Documents Alfa Aesar™
H66267MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6626703 View Documents Alfa Aesar™
H6626703
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1016859-62-3
201.247
4-3-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-3-yl thiazol-2-yl acetonitrile, 2-4-pyridin-3-yl-1,3-thiazol-2-yl acetonitrile
2-(4-pyridin-3-yl-1,3-thiazol-2-yl)acetonitrile
C10H7N3S
FAUDLOFDWKFOQO-UHFFFAOYSA-N
24704442
C1=CC(=CN=C1)C2=CSC(=N2)CC#N

Specifications

Brown
1016859-62-3
UN3439
FAUDLOFDWKFOQO-UHFFFAOYSA-N
2-(4-pyridin-3-yl-1,3-thiazol-2-yl)acetonitrile
24704442
Solid
4-(3-Pyridyl)-2-thiazoleacetonitrile
250mg
C10H7N3S
4-3-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-3-yl thiazol-2-yl acetonitrile, 2-4-pyridin-3-yl-1,3-thiazol-2-yl acetonitrile
C1=CC(=CN=C1)C2=CSC(=N2)CC#N
201.247
201.25
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.