Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Piperidinyloxy)benzamide, 96%

$122.05 - $363.14

Chemical Identifiers

Chemical Name or Material 4-(3-Piperidinyloxy)benzamide
CAS 912761-79-6
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD08061104
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5099303

  SDS
alfa aesar™
H5099303
1g Each for $363.14

AAH50993MD

  SDS
alfa aesar™
H50993MD
250mg Each for $122.05
Description & Specifications

Specifications

Formula Weight 220.27
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD08061104
Molecular Formula C12H16N2O2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
Chemical Name or Material 4-(3-Piperidinyloxy)benzamide
Quantity 1g
CAS 912761-79-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.