missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

$90.05 - $326.32

Chemical Identifiers

CAS 857492-98-9
Molecular Formula C16H13NO4
Formula Weight 283.29
MDL Number MFCD00269588
Synonym 4-[(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
Chemical Name or Material 4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5023403 SDS Alfa Aesar™
H5023403
1g Each for $326.32
AAH50234MD SDS Alfa Aesar™
H50234MD
250mg Each for $90.05
Specifications Description & Specifications

Chemical Identifiers

857492-98-9
283.29
4-[(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
C16H13NO4
MFCD00269588
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

Specifications

1g
C16H13NO4
MFCD00269588
4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid
857492-98-9
283.29
4-[(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning