missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid, 95%, Maybridge

Chemical Identifiers

Assay Percent Range 95%
Molecular Formula C10H8N2O3
Formula Weight 204.18
Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC50901EA Fisher Scientific Company
CC50901EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50901CB SDS Fisher Scientific Company
CC50901CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50901DA SDS Fisher Scientific Company
CC50901DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95%
C10H8N2O3
204.18
4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid

Specifications

10g
95%
204.18
851048-56-1
C10H8N2O3
4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid