Please login to your online account to display your discounted pricing

4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid, 95%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CC50901DA

 View more versions of this product

Catalog No. CC50901DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 204.18
Quantity 1g
Molecular Formula C10H8N2O3
CAS 851048-56-1
Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid
Assay Percent Range 95%

Specifications

Chemical Name or Material 4-(3-Methyl-1, 2, 4-oxadiazol-5-yl)benzoic acid
Assay Percent Range 95%
Molecular Formula C10H8N2O3
Formula Weight 204.18