Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Methoxyphenoxy)piperidine, 99%

$86.19 - $86.19

Chemical Identifiers

Chemical Name or Material 4-(3-Methoxyphenoxy)piperidine
CAS 162402-37-1
Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD08061092
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50964MD

  SDS
alfa aesar™
H50964MD
250mg Each for $86.19
Description & Specifications

Specifications

Formula Weight 207.27
Quantity 250mg
MDL Number MFCD08061092
CAS 162402-37-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Chemical Name or Material 4-(3-Methoxyphenoxy)piperidine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12H17NO2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.