missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(3-Isocyanatobenzyl)morpholine, ≥97%, Maybridge

Chemical Identifiers

CAS 166740-66-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H14N2O2·2HCl
Formula Weight 291.18
MDL Number MFCD08060510
Chemical Name or Material 4-(3-Isocyanatobenzyl)morpholine dihydrochloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC40106DA SDS Fisher Scientific Company
CC40106DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

166740-66-5
100.0
C12H14N2O2·2HCl
MFCD08060510
95%
96.5
95%
291.18
4-(3-Isocyanatobenzyl)morpholine dihydrochloride

Specifications

1g
96.5
95%
291.18
Amber Glass Bottle
95%
166740-66-5
100.0
C12H14N2O2·2HCl
MFCD08060510
4-(3-Isocyanatobenzyl)morpholine dihydrochloride

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger