missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(3-Hydroxypropyl)thiomorpholine 1,1-Dioxide, TCI America™

$242.92 - $854.05

Chemical Identifiers

CAS 205194-33-8
Molecular Formula C7H15NO3S
Molecular Weight (g/mol) 193.261
InChI Key VAJSFWFPXNWBLF-UHFFFAOYSA-N
Synonym 3-(1,1-Dioxothiomorpholino)-1-propanol
PubChem CID 11435473
IUPAC Name 3-(1,1-dioxo-1,4-thiazinan-4-yl)propan-1-ol
SMILES C1CS(=O)(=O)CCN1CCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H114125G View Documents TCI America
H114125G
25g Each for $854.05
 
H11415G View Documents TCI America
H11415G
5g Each for $242.92
 
Specifications

Chemical Identifiers

205194-33-8
193.261
3-(1,1-Dioxothiomorpholino)-1-propanol
3-(1,1-dioxo-1,4-thiazinan-4-yl)propan-1-ol
C7H15NO3S
VAJSFWFPXNWBLF-UHFFFAOYSA-N
11435473
C1CS(=O)(=O)CCN1CCCO

Specifications

Undesignated
25g
C7H15NO3S
VAJSFWFPXNWBLF-UHFFFAOYSA-N
3-(1,1-dioxo-1,4-thiazinan-4-yl)propan-1-ol
11435473
Crystal-Powder at 20°C
4-(3-Hydroxypropyl)thiomorpholine 1,1-Dioxide
72°C
205194-33-8
3-(1,1-Dioxothiomorpholino)-1-propanol
C1CS(=O)(=O)CCN1CCCO
193.261
193.26
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No