Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Hydroxy-1-propynyl)benzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 4-(3-Hydroxy-1-propynyl)benzeneboronic acid pinacol ester
Molecular Formula C15H19BO3
Formula Weight 258.12
MDL Number MFCD16294528
Synonym 3-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2-propyn-1-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190103

  SDS
alfa aesar™
H5190103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5190106

  SDS
alfa aesar™
H5190106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.12
Chemical Name or Material 4-(3-Hydroxy-1-propynyl)benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C15H19BO3
MDL Number MFCD16294528
Synonym 3-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2-propyn-1-ol
Quantity 1g