Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Fluorophenyl)-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-(3-Fluorophenyl)-3-thiosemicarbazide
CAS 70619-48-6
Molecular Formula C7H8FN3S
Formula Weight 185.22
MDL Number MFCD00041286
UN Number UN2811
Beilstein 2804712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1165203

  SDS
alfa aesar™
L1165203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1165206

  SDS
alfa aesar™
L1165206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.22
Chemical Name or Material 4-(3-Fluorophenyl)-3-thiosemicarbazide
Molecular Formula C7H8FN3S
MDL Number MFCD00041286
CAS 70619-48-6
UN Number UN2811
Beilstein 2804712
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 97%
Melting Point ≈160°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.