missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Fluorophenyl)-3-thiosemicarbazide, 97%

Manufacturer:  Alfa Aesar™ L1165206

 View more versions of this product

Catalog No. AAL1165206



Specifications

Specifications

4-(3-Fluorophenyl)-3-thiosemicarbazide
5g
C7H8FN3S
UN2811
n-3-fluorophenyl hydrazinecarbothioamide, 4-3-fluorophenyl-3-thiosemicarbazide, 3-amino-1-3-fluorophenyl thiourea, 1-amino-3-3-fluorophenyl thiourea, 3-fluorophenyl amino hydrazinomethane-1-thione, 4-m-fluorophenyl thiosemicarbazide, 1-azanyl-3-3-fluorophenyl thiourea, n-3-fluorophenyl-1-hydrazinecarbothioamide
C1=CC(=CC(=C1)F)NC(=S)NN
185.22
185.22
~160°C
70619-48-6
MFCD00041286
2804712
HLLFMTUWRLDCPF-UHFFFAOYSA-N
1-amino-3-(3-fluorophenyl)thiourea
2737499
97%

Chemical Identifiers

70619-48-6
185.22
HLLFMTUWRLDCPF-UHFFFAOYSA-N
2737499
C1=CC(=CC(=C1)F)NC(=S)NN
C7H8FN3S
MFCD00041286
n-3-fluorophenyl hydrazinecarbothioamide, 4-3-fluorophenyl-3-thiosemicarbazide, 3-amino-1-3-fluorophenyl thiourea, 1-amino-3-3-fluorophenyl thiourea, 3-fluorophenyl amino hydrazinomethane-1-thione, 4-m-fluorophenyl thiosemicarbazide, 1-azanyl-3-3-fluorophenyl thiourea, n-3-fluorophenyl-1-hydrazinecarbothioamide
1-amino-3-(3-fluorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents