Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 95%

$102.31 - $357.11

Chemical Identifiers

Chemical Name or Material 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
CAS 912761-48-9
Molecular Formula C9H15N3O
Formula Weight 181.24
MDL Number MFCD08061039
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5087403

  SDS
alfa aesar™
H5087403
1g Each for $357.11

AAH50874MD

  SDS
alfa aesar™
H50874MD
250mg Each for $102.31
Description & Specifications

Specifications

Formula Weight 181.24
Molecular Formula C9H15N3O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Chemical Name or Material 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
MDL Number MFCD08061039
Quantity 1g
Health Hazard 1 Warning
CAS 912761-48-9

Applications
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.