Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester, 98%

$54.24 - $159.72

Chemical Identifiers

Chemical Name or Material 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester
CAS 850411-14-2
Molecular Formula C21H30BNO5
Formula Weight 387.28
MDL Number MFCD06657751
Synonym Ethyl 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2808606

  SDS
alfa aesar™
H2808606
5g Each for $159.72

AAH2808603

  SDS
alfa aesar™
H2808603
1g Each for $54.24
Description & Specifications

Specifications

Formula Weight 387.28
Chemical Name or Material 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester
Assay 98%
Molecular Formula C21H30BNO5
MDL Number MFCD06657751
CAS 850411-14-2
Synonym Ethyl 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperidine-3-carboxylate
Quantity 5g