Please login to your online account to display your discounted pricing

{4-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material {4-[3-(Dimethylamino)propoxy]phenyl}methanol
CAS 426831-08-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H19NO2
Formula Weight 209.29
MDL Number MFCD06202771
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC59509DA

  SDS
fisher scientific company
CC59509DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 209.29
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material {4-[3-(Dimethylamino)propoxy]phenyl}methanol
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD06202771
Molecular Formula C12H19NO2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 426831-08-5
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.