missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[3-(Dimethylamino)propoxy]benzylamine, 97%, Maybridge

Chemical Identifiers

CAS 91637-76-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H20N2O
Formula Weight 208.3
MDL Number MFCD07786701
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]benzylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59513CB SDS Fisher Scientific Company
CC59513CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

91637-76-2
100.0
C12H20N2O
MFCD07786701
97%
96.5
97%
208.3
4-[3-(Dimethylamino)propoxy]benzylamine

Specifications

208.3
100.0
97%
96.5
91637-76-2
97%
250mg
C12H20N2O
MFCD07786701
Amber Glass Bottle
4-[3-(Dimethylamino)propoxy]benzylamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger