Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Chlorophenyl)-3-thiosemicarbazide, 98+%

Chemical Identifiers

Chemical Name or Material 4-(3-Chlorophenyl)-3-thiosemicarbazide
CAS 42135-76-2
Molecular Formula C7H8ClN3S
Formula Weight 201.68
MDL Number MFCD00041287
UN Number UN2811
Beilstein 2804713
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1161003

  SDS
alfa aesar™
L1161003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1161006

  SDS
alfa aesar™
L1161006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.68
Chemical Name or Material 4-(3-Chlorophenyl)-3-thiosemicarbazide
MDL Number MFCD00041287
CAS 42135-76-2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay ≥98%
Molecular Formula C7H8ClN3S
UN Number UN2811
Beilstein 2804713
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 130° to 133°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.