missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Chlorophenyl)-3-thiosemicarbazide, 98+%

Manufacturer:  Alfa Aesar™ L1161003

 View more versions of this product

Catalog No. AAL1161003



Specifications

Specifications

4-(3-Chlorophenyl)-3-thiosemicarbazide
1g
C7H8ClN3S
UN2811
4-3-chlorophenyl-3-thiosemicarbazide, 4-3-chlorophenyl thiosemicarbazide, n-3-chlorophenyl hydrazinecarbothioamide, 4-m-chlorophenyl thiosemicarbazide, 1-amino-3-3-chlorophenyl thiourea, 3-amino-1-3-chlorophenyl thiourea, 1-azanyl-3-3-chlorophenyl thiourea, 4-cptsc, 3-chlorophenyl amino hydrazinomethane-1-thione, acmc-1arqr
C1=CC(=CC(=C1)Cl)NC(=S)NN
201.672
201.68
130°C to 133°C
42135-76-2
MFCD00041287
2804713
YUSLJYYTYKWSHL-UHFFFAOYSA-N
1-amino-3-(3-chlorophenyl)thiourea
707050
≥98%

Chemical Identifiers

42135-76-2
201.672
YUSLJYYTYKWSHL-UHFFFAOYSA-N
707050
C1=CC(=CC(=C1)Cl)NC(=S)NN
C7H8ClN3S
MFCD00041287
4-3-chlorophenyl-3-thiosemicarbazide, 4-3-chlorophenyl thiosemicarbazide, n-3-chlorophenyl hydrazinecarbothioamide, 4-m-chlorophenyl thiosemicarbazide, 1-amino-3-3-chlorophenyl thiourea, 3-amino-1-3-chlorophenyl thiourea, 1-azanyl-3-3-chlorophenyl thiourea, 4-cptsc, 3-chlorophenyl amino hydrazinomethane-1-thione, acmc-1arqr
1-amino-3-(3-chlorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents