Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Chlorophenyl)-1-piperazineacetamidoxime

$138.54 - $606.47

Chemical Identifiers

Chemical Name or Material 4-(3-Chlorophenyl)-1-piperazineacetamidoxime
CAS 261623-65-8
Molecular Formula C12H17ClN4O
Formula Weight 268.75
MDL Number MFCD08060957
Synonym 4-(3-Chlorophenyl)-N-hydroxy-1-piperazineacetamidine; 2-[4-(3-Chlorophenyl)piperazin-1-yl]-N-hydroxyethanimidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5067103

 
alfa aesar™
H5067103
1g Each for $138.54

AAH5067106

 
alfa aesar™
H5067106
5g Each for $606.47
Description & Specifications

Specifications

Formula Weight 268.75
Chemical Name or Material 4-(3-Chlorophenyl)-1-piperazineacetamidoxime
Molecular Formula C12H17ClN4O
MDL Number MFCD08060957
CAS 261623-65-8
Synonym 4-(3-Chlorophenyl)-N-hydroxy-1-piperazineacetamidine; 2-[4-(3-Chlorophenyl)piperazin-1-yl]-N-hydroxyethanimidamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.