Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Chlorophenoxy)piperidine, 98%

$112.86 - $112.86

Chemical Identifiers

Chemical Name or Material 4-(3-Chlorophenoxy)piperidine
CAS 97840-40-9
Molecular Formula C11H14ClNO
Formula Weight 211.69
MDL Number MFCD08061087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50956MD

  SDS
alfa aesar™
H50956MD
250mg Each for $112.86
Description & Specifications

Specifications

Formula Weight 211.69
MDL Number MFCD08061087
Quantity 250mg
Assay 98%
Chemical Name or Material 4-(3-Chlorophenoxy)piperidine
Health Hazard 1 Warning
Molecular Formula C11H14ClNO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 97840-40-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.