Learn More
4-{[3-(chloromethyl)isoxazol-5-yl]methyl}morpholine, Tech., Thermo Scientific™
Chemical Identifiers
CAS | 953408-97-4 |
---|---|
Molecular Formula | C9H13ClN2O2 |
Molecular Weight (g/mol) | 216.665 |
MDL Number | MFCD11841078 |
InChI Key | FZRKHZMWHFBKDV-UHFFFAOYSA-N |
Synonym | 3-chloromethyl-5-morpholin-4-yl methyl isoxazole, 4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine |
PubChem CID | 43811054 |
IUPAC Name | 4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine |
SMILES | C1COCCN1CC2=CC(=NO2)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC70824CB
|
Fisher Scientific Company
CC70824CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC70824DA
|
Fisher Scientific Company
CC70824DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
953408-97-4 | |
216.665 | |
FZRKHZMWHFBKDV-UHFFFAOYSA-N | |
43811054 | |
C1COCCN1CC2=CC(=NO2)CCl |
C9H13ClN2O2 | |
MFCD11841078 | |
3-chloromethyl-5-morpholin-4-yl methyl isoxazole, 4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine | |
4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine |
Specifications
953408-97-4 | |
100.0 | |
MFCD11841078 | |
3-chloromethyl-5-morpholin-4-yl methyl isoxazole, 4-3-chloromethyl isoxazol-5-yl methyl morpholine, 4-3-chloromethyl-1,2-oxazol-5-yl methyl morpholine | |
FZRKHZMWHFBKDV-UHFFFAOYSA-N | |
4-[[3-(chloromethyl)-1,2-oxazol-5-yl]methyl]morpholine | |
43811054 | |
Tech. | |
4-{[3-(Chloromethyl)isoxazol-5-yl]methyl}morpholine |
80.0 | |
C9H13ClN2O2 | |
Amber Glass Bottle | |
250mg | |
C1COCCN1CC2=CC(=NO2)CCl | |
216.665 | |
216.67 | |
Technical |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning