missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54229MD

 View more versions of this product

Catalog No. AAH54229MD



Description

Description

Applications
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline
250mg
C21H14ClFIN3O
4-3-chloro-4-3-fluorobenzyloxy phenylamino-6-iodoquinazoline
UHFPFDMMKYQMLC-UHFFFAOYSA-N
N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-iodoquinazolin-4-amine
10174519
97%
Light sensitive
231278-20-9
MFCD09998827
Slightly soluble in water.
C1=CC(=CC(=C1)F)COC2=C(C=C(C=C2)NC3=NC=NC4=C3C=C(C=C4)I)Cl
505.716
505.72

Chemical Identifiers

231278-20-9
505.716
UHFPFDMMKYQMLC-UHFFFAOYSA-N
10174519
C1=CC(=CC(=C1)F)COC2=C(C=C(C=C2)NC3=NC=NC4=C3C=C(C=C4)I)Cl
C21H14ClFIN3O
MFCD09998827
4-3-chloro-4-3-fluorobenzyloxy phenylamino-6-iodoquinazoline
N-[3-chloro-4-[(3-fluorophenyl)methoxy]phenyl]-6-iodoquinazolin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents