Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Bromo-5-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(3-Bromo-5-fluorobenzyl)morpholine
CAS 281652-25-3
Formula Weight 274.13
MDL Number MFCD01307672
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6267303

 
alfa aesar™
H6267303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6267306

 
alfa aesar™
H6267306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62673MD

 
alfa aesar™
H62673MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Physical Form Solid
Quantity 1g
MDL Number MFCD01307672
CAS 281652-25-3
Chemical Name or Material 4-(3-Bromo-5-fluorobenzyl)morpholine
Assay 96%