missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Bromo-5-fluorobenzyl)morpholine, 96%

Manufacturer:  Alfa Aesar™ H6267306

 View more versions of this product

Catalog No. AAH6267306



Specifications

Specifications

4-(3-Bromo-5-fluorobenzyl)morpholine
281652-25-3
MFCD01307672
KRXZMNYOZJCBKI-UHFFFAOYSA-N
4-[(3-bromo-4-fluorophenyl)methyl]morpholine
782859
Solid
5g
C11H13BrFNO
4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905, 2-fluoro-5-morpholin-4-ylmethyl-bromobenzene
C1COCCN1CC2=CC(=C(C=C2)F)Br
274.133
274.13
96%

Chemical Identifiers

281652-25-3
274.133
KRXZMNYOZJCBKI-UHFFFAOYSA-N
782859
C1COCCN1CC2=CC(=C(C=C2)F)Br
C11H13BrFNO
MFCD01307672
4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905, 2-fluoro-5-morpholin-4-ylmethyl-bromobenzene
4-[(3-bromo-4-fluorophenyl)methyl]morpholine
Documents