missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Bromo-4-fluorobenzyl)morpholine, 96%

$70.39 - $837.35

Chemical Identifiers

CAS 281652-25-3
Molecular Formula C11H13BrFNO
Molecular Weight (g/mol) 274.133
MDL Number MFCD01307672
InChI Key KRXZMNYOZJCBKI-UHFFFAOYSA-N
Synonym 2-fluoro-5-morpholin-4-ylmethyl-bromobenzene, 4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905
PubChem CID 782859
IUPAC Name 4-[(3-bromo-4-fluorophenyl)methyl]morpholine
SMILES C1COCCN1CC2=CC(=C(C=C2)F)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62455MD
View Documents
Alfa Aesar™
H62455MD
250mg Each for $70.39
 
AAH6245503
View Documents
Alfa Aesar™
H6245503
1g Each for $208.93
 
AAH6245506
View Documents
Alfa Aesar™
H6245506
5g Each for $837.35
 
Specifications

Chemical Identifiers

281652-25-3
274.133
KRXZMNYOZJCBKI-UHFFFAOYSA-N
782859
C1COCCN1CC2=CC(=C(C=C2)F)Br
C11H13BrFNO
MFCD01307672
2-fluoro-5-morpholin-4-ylmethyl-bromobenzene, 4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905
4-[(3-bromo-4-fluorophenyl)methyl]morpholine

Specifications

250mg
C11H13BrFNO
2-fluoro-5-morpholin-4-ylmethyl-bromobenzene, 4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905
C1COCCN1CC2=CC(=C(C=C2)F)Br
274.133
274.13
96%
281652-25-3
MFCD01307672
KRXZMNYOZJCBKI-UHFFFAOYSA-N
4-[(3-bromo-4-fluorophenyl)methyl]morpholine
782859
Solid
4-(3-Bromo-4-fluorobenzyl)morpholine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures