Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3-Bromo-2-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(3-Bromo-2-fluorobenzyl)morpholine
Formula Weight 274.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6229803

 
alfa aesar™
H6229803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6229806

 
alfa aesar™
H6229806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62298MD

 
alfa aesar™
H62298MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Chemical Name or Material 4-(3-Bromo-2-fluorobenzyl)morpholine
Physical Form Solid
Assay 96%
Quantity 1g