Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(3,5-Dimethyl-4H-1,2,4-triazol-4-yl)piperidine, 95%

$115.45 - $400.22

Chemical Identifiers

Chemical Name or Material 4-(3,5-Dimethyl-4H-1,2,4-triazol-4-yl)piperidine
CAS 795310-52-0
Molecular Formula C9H16N4
Formula Weight 180.25
MDL Number MFCD08061013
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5082303

 
alfa aesar™
H5082303
1g Each for $400.22

AAH50823MD

 
alfa aesar™
H50823MD
250mg Each for $115.45
Description & Specifications

Specifications

Formula Weight 180.25
Chemical Name or Material 4-(3,5-Dimethyl-4H-1,2,4-triazol-4-yl)piperidine
Assay 95%
Molecular Formula C9H16N4
MDL Number MFCD08061013
CAS 795310-52-0
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.