Please login to your online account to display your discounted pricing

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzenesulfonyl chloride, ≥90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC68203DA

 View more versions of this product

Catalog No. CC68203DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 270.74
Quantity 1g
Assay 90%
CAS Max % 100.0
Molecular Formula C11H11ClN2O2S
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
CAS Min % 89.5
MDL Number MFCD09817557
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzenesulfonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 90+%
CAS 61320-20-5
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzenesulfonyl chloride
CAS 61320-20-5
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90+%
Molecular Formula C11H11ClN2O2S
Formula Weight 270.74
MDL Number MFCD09817557
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents