Please login to your online account to display your discounted pricing

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde
CAS 934570-54-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H12N2O
Formula Weight 200.24
MDL Number MFCD06800625
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC68204DA

  SDS
fisher scientific company
CC68204DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.24
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde
Molecular Formula C12H12N2O
CAS 934570-54-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS Min % 96.5
MDL Number MFCD06800625
Packaging Amber Glass Bottle
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.