Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[3-(4-Morpholinyl)propyl]-3-thiosemicarbazide, 98+%

$88.15 - $88.15

Chemical Identifiers

Chemical Name or Material 4-[3-(4-Morpholinyl)propyl]-3-thiosemicarbazide
CAS 32813-48-2
Molecular Formula C8H18N4OS
Formula Weight 218.32
MDL Number MFCD00023400
Beilstein 881684
Synonym 4-[3-(4-Morpholino)propyl]-3-thiosemicarbazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1071403

  SDS
alfa aesar™
L1071403
1g Each for $88.15
Description & Specifications

Specifications

Formula Weight 218.32
Chemical Name or Material 4-[3-(4-Morpholinyl)propyl]-3-thiosemicarbazide
Health Hazard 1 Danger
Beilstein 881684
MDL Number MFCD00023400
Molecular Formula C8H18N4OS
Melting Point 135° to 137°C
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-[3-(4-Morpholino)propyl]-3-thiosemicarbazide
Assay ≥98%
CAS 32813-48-2
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.