missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC33839DA
Specifications
4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine | |
90% | |
C16H24BNO3 | |
4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCOCC3 | |
289.182 | |
289.18 |
852227-95-3 | |
Amber Glass Bottle | |
1g | |
NCJDKFFODGZRRL-UHFFFAOYSA-N | |
4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine | |
4192663 | |
90% |
Chemical Identifiers
852227-95-3 | |
289.182 | |
4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-morpholinophenylboronic acid pinacol ester, 3-morpholino phenylboronic acid pinacol ester, 3-4-morpholino phenylboronic acid pinacol ester, 4-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, pubchem19450, ksc496k1f, 3-morpholinophenylboronic acidpinacolester, 3-morpholinophenylboronic acid pinacol | |
4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine |
C16H24BNO3 | |
NCJDKFFODGZRRL-UHFFFAOYSA-N | |
4192663 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3CCOCC3 |