Please login to your online account to display your discounted pricing

4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 90%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC33839CB

 View more versions of this product

Catalog No. CC33839CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 289.18
Quantity 250mg
Assay 90%
Assay Percent Range 90%
Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Molecular Formula C16H24BNO3
CAS 852227-95-3
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
Assay 90%
Assay Percent Range 90%
Molecular Formula C16H24BNO3
Formula Weight 289.18