Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester

$79.73 - $320.18

Chemical Identifiers

Chemical Name or Material 4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester
CAS 1257648-78-4
Molecular Formula C19H28BN3O2
Formula Weight 341.26
MDL Number MFCD12546046
Synonym 4-[3-(1-Imidazolyl)propylaminomethyl]phenylboronic acid pinacol ester; N-[3-(1-Imidazolyl)propyl)]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5167103

 
alfa aesar™
H5167103
1g Each for $79.73

AAH5167106

 
alfa aesar™
H5167106
5g Each for $320.18
Description & Specifications

Specifications

Formula Weight 341.26
Chemical Name or Material 4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester
CAS 1257648-78-4
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C19H28BN3O2
MDL Number MFCD12546046
Synonym 4-[3-(1-Imidazolyl)propylaminomethyl]phenylboronic acid pinacol ester; N-[3-(1-Imidazolyl)propyl)]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.