Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[2-(Trifluoromethyl)phenoxy]piperidine, 99%

$116.92 - $406.82

Chemical Identifiers

Chemical Name or Material 4-[2-(Trifluoromethyl)phenoxy]piperidine
CAS 824390-04-7
Molecular Formula C12H14F3NO
Formula Weight 245.25
MDL Number MFCD06248881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5081303

  SDS
alfa aesar™
H5081303
1g Each for $406.82

AAH50813MD

  SDS
alfa aesar™
H50813MD
250mg Each for $116.92
Description & Specifications

Specifications

Formula Weight 245.25
MDL Number MFCD06248881
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-[2-(Trifluoromethyl)phenoxy]piperidine
Assay 99%
Molecular Formula C12H14F3NO
CAS 824390-04-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.