missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone, TCI America™

Manufacturer:  TCI America P23705G

 View more versions of this product

Catalog No. P23705G



Specifications

Specifications

4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone
44°C
121040-08-2
MFCD14636435
ISFMATRVDNBZMP-UHFFFAOYSA-N
4-[2-(4-pentylcyclohexyl)ethyl]cyclohexan-1-one
12030409
Crystal-Lump at 20°C
White
5g
C19H34O
4-[2-(trans-4-Amylcyclohexyl)ethyl]cyclohexanone
CCCCCC1CCC(CC1)CCC2CCC(=O)CC2
278.48
278.48
≥98.0% (GC)

Chemical Identifiers

121040-08-2
278.48
ISFMATRVDNBZMP-UHFFFAOYSA-N
12030409
CCCCCC1CCC(CC1)CCC2CCC(=O)CC2
C19H34O
MFCD14636435
4-[2-(trans-4-Amylcyclohexyl)ethyl]cyclohexanone
4-[2-(4-pentylcyclohexyl)ethyl]cyclohexan-1-one
Safety and Handling

Safety and Handling

TSCA : No

Documents