Please login to your online account to display your discounted pricing

4-(2-Thienyl)benzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(2-Thienyl)benzoic acid
CAS 29886-62-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8O2S
Formula Weight 204.25
MDL Number MFCD00690742
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12001DE

 
fisher scientific company
CC12001DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.25
Quantity 5g
Assay 97%
CAS Max % 100.0
Health Hazard 1 Warning
CAS 29886-62-2
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
CAS Min % 96.5
MDL Number MFCD00690742
Assay Percent Range 97%
Chemical Name or Material 4-(2-Thienyl)benzoic acid
Molecular Formula C11H8O2S
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.