Please login to your online account to display your discounted pricing

4-(2-Thienyl)benzenesulfonyl chloride, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride
Assay Percent Range 95%
Molecular Formula C10H7ClO2S2
Formula Weight 258.75
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC451160050

  SDS
thermo fisher scientific
AC451160050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC451160010

  SDS
thermo fisher scientific
AC451160010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.75
Quantity 5g
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride
Molecular Formula C10H7ClO2S2
CAS 209592-91-6
Assay Percent Range 95%