missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Thienyl)benzaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 107834-03-7
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.24
MDL Number MFCD02682003
InChI Key UECDQUOWFRTJOH-UHFFFAOYSA-N
Synonym 2-4-formylphenyl thiophene, 4-2-thienyl benzaldehyde, 4-2-thienyl benzaldeyde, 4-thien-2-yl benzaldehyde, 4-thiophen-2-yl benzaldehyde, 4-thiophen-2-yl-benzaldehyde, acmc-209vfl, benzaldehyde, 4-2-thienyl, benzaldehyde,4-2-thienyl
PubChem CID 736499
IUPAC Name 4-thiophen-2-ylbenzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CS1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC12004DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC12004DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

107834-03-7
188.24
UECDQUOWFRTJOH-UHFFFAOYSA-N
736499
O=CC1=CC=C(C=C1)C1=CC=CS1
C11H8OS
MFCD02682003
2-4-formylphenyl thiophene, 4-2-thienyl benzaldehyde, 4-2-thienyl benzaldeyde, 4-thien-2-yl benzaldehyde, 4-thiophen-2-yl benzaldehyde, 4-thiophen-2-yl-benzaldehyde, acmc-209vfl, benzaldehyde, 4-2-thienyl, benzaldehyde,4-2-thienyl
4-thiophen-2-ylbenzaldehyde

Specifications

Amber Glass Bottle
107834-03-7
100.0
C11H8OS
2-4-formylphenyl thiophene, 4-2-thienyl benzaldehyde, 4-2-thienyl benzaldeyde, 4-thien-2-yl benzaldehyde, 4-thiophen-2-yl benzaldehyde, 4-thiophen-2-yl-benzaldehyde, acmc-209vfl, benzaldehyde, 4-2-thienyl, benzaldehyde,4-2-thienyl
O=CC1=CC=C(C=C1)C1=CC=CS1
188.24
188.25
4-(2-Thienyl)benzaldehyde
1g
96.5
97%
MFCD02682003
UECDQUOWFRTJOH-UHFFFAOYSA-N
4-thiophen-2-ylbenzaldehyde
736499
97%
SDS
Product Certifications