missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Thienyl)aniline, 97%

$169.04 - $169.04

Chemical Identifiers

CAS 70010-48-9
Molecular Formula C10H9NS
Formula Weight 175.25
MDL Number MFCD01224064
Synonym 2-(4-Aminophenyl)thiophene
Chemical Name or Material 4-(2-Thienyl)aniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2775303 SDS Alfa Aesar™
H2775303
1g Each for $169.04
Specifications Description & Specifications

Chemical Identifiers

70010-48-9
175.25
2-(4-Aminophenyl)thiophene
C10H9NS
MFCD01224064
4-(2-Thienyl)aniline

Specifications

74° to 78°C
70010-48-9
175.25
2-(4-Aminophenyl)thiophene
97%
1g
C10H9NS
MFCD01224064
4-(2-Thienyl)aniline

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger