missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Thienyl)-3-buten-2-one, 97%

$41.28 - $531.05

Chemical Identifiers

CAS 874-83-9
Molecular Formula C8H8OS
Formula Weight 152.22
MDL Number MFCD00053051
Chemical Name or Material 4-(2-Thienyl)-3-buten-2-one
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2519103 SDS Alfa Aesar™
B2519103
1g Each for $41.28
AAB2519106 SDS Alfa Aesar™
B2519106
5g Each for $135.53
AAB2519114 SDS Alfa Aesar™
B2519114
25g Each for $531.05
Specifications Description & Specifications

Chemical Identifiers

874-83-9
152.22
4-(2-Thienyl)-3-buten-2-one
C8H8OS
MFCD00053051

Specifications

86° to 88°C (1mmHg)
1g
C8H8OS
MFCD00053051
97%
22° to 25°C
874-83-9
152.22
4-(2-Thienyl)-3-buten-2-one