Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 96%

$36.18 - $36.18

Chemical Identifiers

Chemical Name or Material 4-(2-Tetrahydropyranyloxy)benzeneboronic acid
CAS 182281-01-2
Molecular Formula C11H15BO4
Formula Weight 222.05
MDL Number MFCD02183525
Synonym 4-(2-Tetrahydropyranyloxy)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5044503

 
alfa aesar™
H5044503
1g Each for $36.18
Description & Specifications

Specifications

Formula Weight 222.05
Chemical Name or Material 4-(2-Tetrahydropyranyloxy)benzeneboronic acid
Assay 96%
Molecular Formula C11H15BO4
MDL Number MFCD02183525
CAS 182281-01-2
Synonym 4-(2-Tetrahydropyranyloxy)phenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C