Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

$30.74 - $30.74

Chemical Identifiers

Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
CAS 16593-81-0
Molecular Formula C11H8N3NaO2·H2O
Formula Weight 255.21 (237.19 Anhydrous)
MDL Number MFCD00150712
Beilstein 3825637
Synonym PAR-Na
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1810303

  SDS
alfa aesar™
A1810303
1g Each for $30.74
Description & Specifications

Specifications

Formula Weight 255.21 (237.19 Anhydrous)
Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
Synonym PAR-Na
Quantity 1g
MDL Number MFCD00150712
Melting Point >300°C
Health Hazard 1 Warning
CAS 16593-81-0
Beilstein 3825637
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H8N3NaO2·H2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.