missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Pyridyl)benzaldehyde, TCI America™

Manufacturer:  TCI America P16715G

 View more versions of this product

Catalog No. P16715G



Specifications

Specifications

4-(2-Pyridyl)benzaldehyde
50°C
127406-56-8
MFCD01863537
NMLYGLCBSFKJFI-UHFFFAOYSA-N
4-pyridin-2-ylbenzaldehyde
1515240
Crystal-Powder at 20°C
Yellow
5g
C12H9NO
4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
C1=CC=NC(=C1)C2=CC=C(C=C2)C=O
183.21
183.21
≥95.0% (GC,T)

Chemical Identifiers

127406-56-8
183.21
NMLYGLCBSFKJFI-UHFFFAOYSA-N
1515240
C1=CC=NC(=C1)C2=CC=C(C=C2)C=O
C12H9NO
MFCD01863537
4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde
4-pyridin-2-ylbenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents