missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(2-Pyridyl)benzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5549803
Description
4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(2-Pyridyl)benzaldehyde is used as a reactive metabolite of Atazanavir
Solubility
Soluble in Chloroform
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
4-(2-Pyridyl)benzaldehyde | |
50°C to 52°C | |
C12H9NO | |
1 g | |
Soluble in Chloroform | |
O=CC1=CC=C(C=C1)C1=CC=CC=N1 | |
183.21 | |
183.21 |
127406-56-8 | |
113°C (235°F) | |
MFCD01863537 | |
4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde | |
NMLYGLCBSFKJFI-UHFFFAOYSA-N | |
4-pyridin-2-ylbenzaldehyde | |
1515240 | |
98% |
Chemical Identifiers
127406-56-8 | |
183.21 | |
NMLYGLCBSFKJFI-UHFFFAOYSA-N | |
1515240 | |
O=CC1=CC=C(C=C1)C1=CC=CC=N1 |
C12H9NO | |
MFCD01863537 | |
4-2-pyridyl benzaldehyde, 4-2-pyridinyl benzaldehyde, 4-pyridin-2-yl benzaldehyde, 4-pyridin-2-yl-benzaldehyde, 2-4-formylphenyl pyridine, 4-2-pyridinyl-benzaldehyde, benzaldehyde, 4-2-pyridinyl, 4-2-pyridyl-benzaldehyde, pubchem18779, 2-pyridyl benzaldehyde | |
4-pyridin-2-ylbenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only