missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Pyridyl)-2-methyl-3-butyn-2-ol, 98+%

$58.99 - $188.04

Chemical Identifiers

CAS 29767-97-3
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00168867
InChI Key YBUWIVMOMHEALF-UHFFFAOYSA-N
Synonym 2-methyl-4-2-pyridinyl-3-butyn-2-ol
PubChem CID 698810
IUPAC Name 2-methyl-4-pyridin-2-ylbut-3-yn-2-ol
SMILES CC(C)(C#CC1=CC=CC=N1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1767603 View Documents Alfa Aesar™
L1767603
1g Each for $58.99
 
AAL1767606 View Documents Alfa Aesar™
L1767606
5g Each for $188.04
 
Specifications

Chemical Identifiers

29767-97-3
161.204
YBUWIVMOMHEALF-UHFFFAOYSA-N
698810
CC(C)(C#CC1=CC=CC=N1)O
C10H11NO
MFCD00168867
2-methyl-4-2-pyridinyl-3-butyn-2-ol
2-methyl-4-pyridin-2-ylbut-3-yn-2-ol

Specifications

120°C (1.5mmHg)
1g
C10H11NO
1447913
YBUWIVMOMHEALF-UHFFFAOYSA-N
2-methyl-4-pyridin-2-ylbut-3-yn-2-ol
698810
≥98%
80°C to 85°C
29767-97-3
MFCD00168867
2-methyl-4-2-pyridinyl-3-butyn-2-ol
CC(C)(C#CC1=CC=CC=N1)O
161.204
161.2
4-(2-Pyridyl)-2-methyl-3-butyn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures