missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(2-Pyridyl)-2-methyl-3-butyn-2-ol, 98+%

Manufacturer:  Alfa Aesar™ L1767606

 View more versions of this product

Catalog No. AAL1767606



Specifications

Specifications

4-(2-Pyridyl)-2-methyl-3-butyn-2-ol
80°C to 85°C
29767-97-3
MFCD00168867
2-methyl-4-2-pyridinyl-3-butyn-2-ol
CC(C)(C#CC1=CC=CC=N1)O
161.204
161.2
120°C (1.5mmHg)
5g
C10H11NO
1447913
YBUWIVMOMHEALF-UHFFFAOYSA-N
2-methyl-4-pyridin-2-ylbut-3-yn-2-ol
698810
≥98%

Chemical Identifiers

29767-97-3
161.204
YBUWIVMOMHEALF-UHFFFAOYSA-N
698810
CC(C)(C#CC1=CC=CC=N1)O
C10H11NO
MFCD00168867
2-methyl-4-2-pyridinyl-3-butyn-2-ol
2-methyl-4-pyridin-2-ylbut-3-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents