missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

$39.25 - $114.61

Chemical Identifiers

CAS 918655-03-5
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD09260454
InChI Key ICQAKBYFBIWELX-UHFFFAOYSA-N
Synonym 4-2-naphthyl benzeneboronic acid, 4-2-naphthyl phenyl boronic acid, 4-2-naphthyl phenylboronic acid, 4-naphth-1-yl benzeneboronic acid, 4-naphthalen-1-yl-boronic acid, 4-naphthalen-2-yl phenyl boronic acid, 4-naphthalen-2-yl phenylboronic acid, 4-naphthalen-2-yl phenylboronicacid
PubChem CID 45382258
IUPAC Name (4-naphthalen-2-ylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0946200MG View Documents Tci America
N0946200MG
200mg Each for $39.25
 
N09461G View Documents Tci America
N09461G
1g Each for $114.61
 
Specifications

Chemical Identifiers

918655-03-5
248.088
ICQAKBYFBIWELX-UHFFFAOYSA-N
45382258
B(C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2)(O)O
C16H13BO2
MFCD09260454
4-2-naphthyl benzeneboronic acid, 4-2-naphthyl phenyl boronic acid, 4-2-naphthyl phenylboronic acid, 4-naphth-1-yl benzeneboronic acid, 4-naphthalen-1-yl-boronic acid, 4-naphthalen-2-yl phenyl boronic acid, 4-naphthalen-2-yl phenylboronic acid, 4-naphthalen-2-yl phenylboronicacid
(4-naphthalen-2-ylphenyl)boronic acid

Specifications

200mg
C16H13BO2
4-2-naphthyl benzeneboronic acid, 4-2-naphthyl phenyl boronic acid, 4-2-naphthyl phenylboronic acid, 4-naphth-1-yl benzeneboronic acid, 4-naphthalen-1-yl-boronic acid, 4-naphthalen-2-yl phenyl boronic acid, 4-naphthalen-2-yl phenylboronic acid, 4-naphthalen-2-yl phenylboronicacid
B(C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2)(O)O
248.088
248.09
4-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride)
918655-03-5
MFCD09260454
ICQAKBYFBIWELX-UHFFFAOYSA-N
(4-naphthalen-2-ylphenyl)boronic acid
45382258
Crystal-Powder at 20°C
Safety and Handling

Safety and Handling

TSCA : No