missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(2-Naphthyl)phenol, 98%, Thermo Scientific™
$369.90 - $1695.90
Chemical Identifiers
CAS | 6336-82-9 |
---|---|
Molecular Formula | C16H12O |
Molecular Weight (g/mol) | 220.27 |
MDL Number | MFCD18312886 |
InChI Key | NIRHUNSXEDESLN-UHFFFAOYSA-N |
Synonym | 4-naphthalen-2-yl phenol, 4-naphthalen-2-yl-phenol, 4-2-naphthyl phenol, d0l3ax, 2-4-hydroxyphenyl naphthalene |
PubChem CID | 236868 |
SMILES | OC1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6433203
|
Thermo Scientific Chemicals
H6433203 |
1 g |
Each for $369.90
|
|
|||||
AAH6433206
|
Thermo Scientific Chemicals
H6433206 |
5 g |
Each for $1,695.90
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6336-82-9 | |
220.27 | |
NIRHUNSXEDESLN-UHFFFAOYSA-N | |
236868 |
C16H12O | |
MFCD18312886 | |
4-naphthalen-2-yl phenol, 4-naphthalen-2-yl-phenol, 4-2-naphthyl phenol, d0l3ax, 2-4-hydroxyphenyl naphthalene | |
OC1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1 |
Specifications
6336-82-9 | |
MFCD18312886 | |
2524079 | |
NIRHUNSXEDESLN-UHFFFAOYSA-N | |
4-(naphthalen-2-yl)phenol | |
236868 | |
98% | |
4-(2-Naphthyl)phenol |
C16H12O | |
1 g | |
4-naphthalen-2-yl phenol, 4-naphthalen-2-yl-phenol, 4-2-naphthyl phenol, d0l3ax, 2-4-hydroxyphenyl naphthalene | |
OC1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1 | |
220.27 | |
220.27 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures