missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Maybridge

Chemical Identifiers

CAS 140836-69-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C13H20N2O2
Formula Weight 236.31
MDL Number MFCD07772849
Chemical Name or Material [4-(2-Morpholinoethoxy)phenyl]methylamine
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC42113DA SDS Fisher Scientific Company
CC42113DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42113CB SDS Fisher Scientific Company
CC42113CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

140836-69-7
100.0
C13H20N2O2
MFCD07772849
95%
94.5
95%
236.31
[4-(2-Morpholinoethoxy)phenyl]methylamine

Specifications

236.31
100.0
C13H20N2O2
94.5
MFCD07772849
95%
1g
140836-69-7
95%
Amber Glass Bottle
[4-(2-Morpholinoethoxy)phenyl]methylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger